Oakton™ Six+ Portable Salinity Meters
For quick, accurate, affordable and single point salinity measurements
Manufacturer:  Oakton™ 3560444
 View more versions of this product
Catalog No. 15-500-108
Includes: SALT 6+ Meter: Conductivity/TDS temperature probe (K= 1), Protective rubber boot, Batteries SALT 6+ Meter Kit: Conductivity/TDS temperature probe (K= 1), Protective rubber boot, Batteries, Calibration solutions and sample bottles, Hard carrying case
Description
Description
SALT 6+ Meter
  • Measures salinity in ppt or %
  • Easy to read large display
  • Convenient push-button operation with hold and auto-off functions
  • Available with manual and automatic modes
  • With protective rubber boot that also acts as meter stand
Specifications
Specifications
Meter, conductivity/TDS temperature probe (K=1), protective rubber boot, calibration solutions, sample bottles, hard carrying case, batteries
±0.5°C
15.7mm
12.7 to 0.12mm
10° to 110°C
Salinity Meter Kit
0.6 in.
5.12 in.
50° to 230°F
0 to 50ppt
Documents