Please login to your online account to display your discounted pricing

Oakton™ Portable Waterproof Colorimeters


Fast, accurate chlorine or chlorine/pH measurements

Manufacturer: oakton™  3564520

 View more versions of this product

Catalog No. 13-300-254

  • / Each
 

Description & Specifications

Specifications

Product Type Free/Total Chlorine colorimeter kit
Accuracy 0.01 to 1.99ppm: 0.01/2 to 6 : 0.1ppm
Display Type LCD
Height (Metric) 4.6cm
Length (English) 6.1 in.
Length (Metric) 15.5cm
Resolution 0.01ppm, 0.1ppm, 1ppm
Weight (English) 7 oz.
Weight (Metric) 200g
Width (English) 2.7 in.
Width (Metric) 6.8cm
IP Rating IP67
Warranty 2 Year
Includes Foil pack reagents (100 free Chlorine tests, 100 total Chlorine tests), sample vials, batteries, hard carrying case and instructions
Operating Temperature Range 0° to 50°C
Wavelength 525nm
Certifications/Compliance US EPA, DPD
Dimensions (L x W x H) 6.1 x 2.7 x 1.8 in. (15.5 x 6.8 x 4.6cm)
Height (English) 1.8 in.