Please login to your online account to display your discounted pricing
Oakton™ Portable Waterproof Colorimeters
Fast, accurate chlorine or chlorine/pH measurements
$842.13 - $842.13
Products
Catalog Number Mfr. No. Price Quantity    
13-300-254
oakton™
3564520
Each for $842.13
Description & Specifications
  • Free chlorine range: 0.01 to 1.99ppm; total chlorine range: 2.0 to 6.0ppm; pH range: 5.9 to 8.2
  • Unique sample chamber uses vial cap as light shield, reducing chance of cover loss
  • Large custom display warns of irregularities from detector or light source
  • 3000 readings on a single set of batteries
  • Easy-to-use pushbutton operation
Both models measure free and total chlorine; Model C301 also measures pH (range: 5.9 to 8.2).
Replacement reagents for Models C301 and C302 are available.
Uses EPA-approved DPD method for chlorine testing