missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oakton™ pHTestr 50S Waterproof Pocket Tester with Spear Tip

Oakton pH 50 Spear Waterproof Pocket pH TestrTM unique junction design provides increased electrolyte flow, resulting in greater contact between sample and reference, ensuring fast, stable readings. Large electrode junction makes clogging virtually impossible

Manufacturer:  Oakton™ 3563425

Catalog No. 11-747-328



Includes: Protective plastic storage case, lanyard, and 4 AAA batteries.

Description

Description

  • Rugged spear tip makes it ideal for soft solids and semisolid materials
  • Easy-to-navigate interface for quick setup and calibration
  • Replaceable, double-junction electrode sensors save time and money
  • Automatic temperature compensation ensures accurate reading in varying environments

Warranty

1 Year

Specifications

Specifications

±0.01
3
Graphics, Dot Matrix 80 x 100 Pixel
0°C to 60°C
250+ hr.
1
Water quality testing, buffer and reagent production, electrochemistry
IP-67
English
Hold Data
Integral electrode
0.01
Checkmark indicator
41°F to 113°F (ambient)
32°F to 140°F
1 Year
Yes
Up to 3 points
pH, Temperature
None
4 AAA alkaline
Four AAA alkaline batteries
Protective plastic storage case, lanyard, and 4 AAA batteries.
Membrane
Valox™
50 Series
-1 to 15
0.1°F/°C
Manual
5°C to 45°C (ambient)
pH Meter
SDS
Documents
Product Certifications
Promotions

Promotions