Please login to your online account to display your discounted pricing

Oakton™ pH and pH/Conductivity Electrode Replacements

$226.77 - $650.63

Specifications

Length (English) 5.12 in.
Length (Metric) 130mm
pH Range 0 to 14
Storage Requirements Oakton electrode storage solution or a 2 to 4mol/L KCl solution
Tip Type Round
View More Specs
Products
Catalog Number Mfr. No. Temperature (English) Operating Connector Types Junction Material Price Quantity    

15-500-012

 
oakton™
3563050
32° to 212°F BNC / Mini-Phone ATC Single ABS Each for $650.63

15-500-016

 
oakton™
3581171
32° to 176°F BNC/ATC Double Epoxy Each for $226.77
Description & Specifications

Specifications

Length (English) 5.12 in.
Length (Metric) 130mm
pH Range 0 to 14
Storage Requirements Oakton electrode storage solution or a 2 to 4mol/L KCl solution
Tip Type Round
Warranty 1 Year