Please login to your online account to display your discounted pricing

Oakton™ pH buffer pouches


OAKTON pH buffer pouches; package of 20 Economical pouches are accurate and convenient.±0.01 accuracy at 77°°F (25°°C)

pH 10.00

Manufacturer: oakton™  WD3565303

 View more versions of this product

Catalog No. 13-300-148

  • / Pack of 20 Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Oakton pH buffer pouches
CAS 877-24-7
pH 10.00

Specifications

Chemical Name or Material Oakton pH buffer pouches
CAS 877-24-7