Oakton™ pH 700, CON 700, pH-CON 700 pH/Conductivity Benchtop Meters
Compact, space-saving design with a larger, easier-to-read display
Manufacturer:  Oakton™ 3541910
 View more versions of this product
Catalog No. 13-620-748
Description
Description
Compact, space-saving design with a larger, easier-to-read display
Specifications
Specifications
ATC
0° to 100°C
0 to 14
±0.01
±0.1 from 199.9mV, ±1 beyond 200mV
Automatic or manual from 0° to 100°C
0.1°C
LCD
Measures pH, mV, and temperature (C or F)
3 Years
32° to 212°F
Meter, double-junction pH electrode, temp. probe, electrode stand, universal power supply/adapters
0.01
±2000mV
±0.2 within 199.9mV, ±2 beyond 200mV
5
±0.3°C
100
120/240V
pH 700
Documents