Oakton™ pH 2700/PC 2700/Ion 2700/CON 2700 Benchtop Multiparameter Meters
Compact benchtop meters to measure pH/mV/Ion/Temperature/Conductivity/TDS/Salinity/Resistivity
Manufacturer:  Oakton™ 3541200
 View more versions of this product
Catalog No. 13-620-453
Includes: pH 2700 and Ion 2700 meter kit includes pH electrode, ATC probe, electrode stand, and software
pH 2700 and Ion 2700 meter includes electrode stand and software
PC 2700 meter kit includes pH electrode, conductivity/temp. probe and electrode stand
CON 2700 meter kit includes 4-cell, K=1 conductivity/temp. probe and electrode stand
Description
Description
Compact benchtop meters to measure pH/mV/Ion/Temperature/Conductivity/TDS/Salinity/Resistivity
Specifications
Specifications
4 Cell, K=1 conductivity/temperature probe and electrode stand
0.01/0.1μS, 0.001/0.01/0.1mS
0.01/0.1ppm/0.001/0.01/0.1ppt
0.01/0.1ppm, 0.001/0.01ppt
0.01megohm
0° to 100°C
±0.5°C
500
CE
15.54cm
110/240V
0μS to 500mS
0 to 500ppt
0 to 80ppt
0 to 20megohm
Up to 6
0.1°C
LCD
RS232
2.75 in.
CON 2700 Meter Kit
17.44cm
Documents