Oakton™ pH 2700/PC 2700/Ion 2700/CON 2700 Benchtop Multiparameter Meters
Compact benchtop meters to measure pH/mV/Ion/Temperature/Conductivity/TDS/Salinity/Resistivity
Manufacturer:  Oakton™ 3542020
 View more versions of this product
Catalog No. 13-620-451
Includes: pH 2700 and Ion 2700 meter kit includes pH electrode, ATC probe, electrode stand, and software
pH 2700 and Ion 2700 meter includes electrode stand and software
PC 2700 meter kit includes pH electrode, conductivity/temp. probe and electrode stand
CON 2700 meter kit includes 4-cell, K=1 conductivity/temp. probe and electrode stand
Description
Description
Compact benchtop meters to measure pH/mV/Ion/Temperature/Conductivity/TDS/Salinity/Resistivity
Specifications
Specifications
Meter, pH electrode, ATC temperature/conductivity probe, 60mL of electrode fill solution, electrode holder, AC adapter and CyberComm software
0.1/0.01/0.001
±2000mV
±0.2mV
0° to 100°C
±0.5°C
500
CE
2.75 in.
pH 2700 Meter Kit
17.44cm
-2 to 20
±0.002
0.1/1mV
Up to 6
0.1°C
LCD
RS232
pH measurement for drinking water, food, pools, laboratory and pharmaceuticals
15.54cm
110/240V
Documents