Oakton™ pH 2700/PC 2700/Ion 2700/CON 2700 Benchtop Multiparameter Meters
Compact benchtop meters to measure pH/mV/Ion/Temperature/Conductivity/TDS/Salinity/Resistivity
Manufacturer:  Oakton™ 3542102
 View more versions of this product
Catalog No. 13-620-450
Includes: pH 2700 and Ion 2700 meter kit includes pH electrode, ATC probe, electrode stand, and software
pH 2700 and Ion 2700 meter includes electrode stand and software
PC 2700 meter kit includes pH electrode, conductivity/temp. probe and electrode stand
CON 2700 meter kit includes 4-cell, K=1 conductivity/temp. probe and electrode stand
Description
Description
Compact benchtop meters to measure pH/mV/Ion/Temperature/Conductivity/TDS/Salinity/Resistivity
Specifications
Specifications
pH electrode, ATC probe, electrode stand, software
0.1/0.01/0.001
±2000mV
±0.2mV
Up to 3 digits
0° to 100°C
±0.5°C
500
CE
2.75 in.
Ion 2700 Meter
17.44cm
-2 to 20
±0.002
0.1/1mV
0 to 19999ppm
Up to 6
0.1°C
LCD
RS232
pH measurement for drinking water, food, pools, laboratory and pharmaceuticals
15.54cm
110/240V
Documents