Oakton™ PC 650 Waterproof Handheld Multiparameter Meter Kit
Oakton™ PC 650 Waterproof Handheld Meter Kit features a top-of-the-line dual parameter meter
Manufacturer:  Oakton™ 3543170
Catalog No. 15-500-057
Description
Description
  • Simultaneously measure your choice of pH, mV, or ISE and conductivity
  • Log up to 500 data points with GLP compliance
  • Wireless IrDA communication for truly waterproof interfacing capabilities
Specifications
Specifications
± 1% full scale
±0.2 mV or 0.05%, whichever is greater
4 x AA
K=0.3
Automatic or manual
Oakton PC 650 Waterproof Handheld Meter Kit
double-junction pH electrode and a 4-electrode (K = 0.3) conductivity cell, pH buffer solutions, conductivity standards, protective rubber boot, data management software, probe holder, hard carrying case, and batteries.
PC 650
0.001 to 19,900 ppm, molar, or mg/L
14°F to 230°F
5.7 cm
LCD
0 to 500 mS/cm
80.00 ppt
±2000 mV
-10°C to +110°C
±0.5% of reading
±0.002
Up to 6 points
CE; IP67
500
18.4 cm
8.3 cm
Multiparameter Meter
0.4 to 1.0
3 Years
1
IP 67
0 to 20.000 Mohm
0 to 500 ppt (TDS factor dependent)
-2.000 to 20.000