missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oakton™ Handheld Ion Meters

Rugged, compact meters with high accuracy

$913.42 - $1,069.07

Specifications

Accuracy (Ion) ±1% FS
Accuracy (mV) ±0.2 within 199.9mV, ±2 beyond 200mV
Accuracy (pH) ±0.01
Accuracy (Temperature) ±0.5°C
Battery Life >200 hr.
View More Specs

Includes: Meter comes with includes protective rubber boot and batteries. Meter Kit includes meter, single-junction, epoxy-body pH electrode, temperature probe, calibration buffer pouches, sample bottles, protective rubber boot, batteries and hard carrying case.


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
13-300-104 Oakton™
3561382
Meter, single-junction, Epoxy body pH electrode, temperature probe, calibration buffer pouches, sample bottles, protective rubber boot, batteries and hard carrying case Each for $1,069.07
13-300-105 Oakton™
3561380
Meter, protective rubber boot and batteries Each for $913.42
Specifications

Description

  • Simple push-button operation
  • Automatic temperature compensation (ATC)
  • Hold and Auto-off functions
  • Built-in stand for benchtop or long-term use
  • Protective boot helps shield meter from drops and dings
  • Meter kits contain everything needed for calibration and measurement and is packaged in a hard carrying case

  • Direct readout of ion concentration
  • Measurement mode: mV
  • Use with pH or most ISE electrodes with BNC connectors (Range: ±500mV)
  • Preprogrammed calibration points
  • pH and temperature (°C) readout with high ±0.01 pH/±0.5°C accuracy

Certifications

ISO 9001:2000, CE marked (Meters)

Warranty

Three years warranty (meter only)

Description & Specifications

Specifications

±1% FS
±0.01
>200 hr.
0 to 14
Simple push-button operation, automatic temperature compensation, hold and auto-off functions, built-in stand for benchtop or long-term use
3.5cm
0.01 to 1999U
0.1 Within ±199.9, 1 beyond ±200mV
Automatic or manual from 0° to 100°C
0 to 50°C
0.9 lb.
2.75 in.
±0.2 within 199.9mV, ±2 beyond 200mV
±0.5°C
Four AAA
0.01
1.37 in.
14cm
0.01U (0.01 to 0.99U), 0.1U (0.1 to 199.9U), 1U (200 to 1999U)
0.1°C
32° to 122°F
0° to 100°C
0.4kg