Please login to your online account to display your discounted pricing

Oakton™ EcoTestr pH

The EcoTestr pH1 and EcoTester pH2 — new and economical quality pH testers designed for fuss-free pH measurements.

$83.05 - $105.30

Specifications

Accuracy ±0.1pH
Accuracy (pH) ±0.1
Battery Life >100 hr.
Battery Type Alkaline
IP Rating 67
View More Specs
Products
Catalog Number Mfr. No. Inputs Range (pH) Price Quantity    

S90520A

 
oakton™
35423-00
EcoTestr pH1 0 to 14 Each for $83.05

S90521A

 
oakton™
35423-10
EcoTestr pH2 -1 to 15 Each for $105.30
Description & Specifications

Specifications

Accuracy ±0.1pH
Accuracy (pH) ±0.1
Battery Life >100 hr.
Battery Type Alkaline
IP Rating 67
Resolution (pH) 0.1
For Use With (Application) Ideal for classroom and field exercises, quick pH checks in hydroponic gardening, environment monitoring, aquariums, aquaculture, agriculture, swimming pools, spa and hot tubs, simple lab work and other water/wastewater applications
Length (Metric) 16.5cm
Resolution 0.1pH
Temperature (English) Operating 32° to 122°F
Temperature (Metric) Operating 0 to 50°C
Temperature (Metric) Range 0° to 50°C
Weight (English) 4.8 oz.
Weight (Metric) 0.13kg
Width (English) 1.5 in.
Width (Metric) 3.8cm

Accurate and easy to use.

  • Full pH Range: 0.0 to 14.0
  • Auto-calibration, auto-buffer recognition and automatic temperature compensation
  • Nonvolatile memory
  • Tester floats on water for easy retrieval
  • Long sensor life — no need to change sensors
  • Click-lock battery compartment requires no tools for battery replacement
  • Hold function freezes reading temporarily for ease of recording
  • Sensor protective cap doubles as a container for samples or calibration solution
  • EcoTestr pH2 offers three-point calibration and automatic temperature compensation (ATC)
  • Auto-Off: 8 minutes, 30 seconds after last key pressed
  • Ingress Rating: IP67 waterproof

Ideal for classroom and field exercises, quick pH checks in hydroponic gardening, environment monitoring, aquariums, aquaculture, agriculture, swimming pools, spa and hot tubs, simple lab work and other water/wastewater applications.