Oakton™ DO Six+ Portable Dissolved Oxygen Meters
No warm up time required for low-maintenance galvanic probe
Manufacturer:  Oakton™ 3564312
 View more versions of this product
Catalog No. 15-500-110
Description
Description
DO 6+ Meter
  • Measures dissolved oxygen in ppm and mg/L
  • Clear LCD with enunciators to aid in use and troubleshooting
  • Six simple buttons allow for freezing readings, toggling between display units and temperature results, setup, calibration, and automatic barometric and salinity compensation after manual parameter inputs
  • Simple design to view electrode slope, zero offset and millivolt values for electrode diagnostics
  • Accurate results with economical pricing
  • Manual input with meter calculated corrections for pressure and salinity
  • With protective rubber boot that also acts as benchtop stand
  • Automatic temperature compensation
For testing water quality in wastewater, surface water, ground water and drinking water
Specifications
Specifications
DO Six+ Portable Dissolved Oxygen Meter
Meter, Galvanic dissolved oxygen probe, electrolyte solution, membrane caps, protective rubber boot, 4 AA batteries
±0.5 °C
AA
±1.5% FS
0 to 20mg/L
0.01mg/L
0.1%
0° to 50°C
3 Years
0.1°C
LCD
CE
0° to 50 °C
0 to 50ppt
0 to 200%
32° to 122°F
Documents