Oakton™ CON 6+ Portable Conductivity Meters
Features five measuring ranges (upto 200mS)
Manufacturer:  Oakton™ 3560400
 View more versions of this product
Catalog No. 15-500-103
Description
Description
Features five measuring ranges (upto 200mS)
Specifications
Specifications
Conductivity Meter
0 to 20μS, 0 to 200μS, 0 to 2000μS, 0 to 20mS, 0 to 200mS
±1% FS
0.1°C
100 hr.
5
1.68 in.
15.7cm
-10° to +110°C
3.37 in.
Meter, conductivity/TDS temp probe, protective rubber boot, batteries
0.01μS, 0.1μS, 1μS, 0.01mS, 0.1mS
Automatic
±0.5°C
Four AA
0.1, 1.0, 10cm-1
4.2cm
Conductivity
1.5V
Documents