missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OAC-1, Tocris™

Oct4 activator; enhances iPSC reprogramming efficiency

$181.19 - $762.58

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C14H11N3O
Formula Weight 237.26
Synonym N-1H-Pyrrolo[2, 3-c]pyridin-5-ylbenzamide
Chemical Name or Material OAC-1
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
488710 R & D Systems
488710
10mg Each for $181.19
488750 R & D Systems
488750
50mg Each for $762.58
Specifications Description & Specifications

Chemical Identifiers

>98%
237.26
OAC-1
C14H11N3O
N-1H-Pyrrolo[2, 3-c]pyridin-5-ylbenzamide

Specifications

10mg
>98%
237.26
OAC-1
300586-90-7
C14H11N3O
N-1H-Pyrrolo[2, 3-c]pyridin-5-ylbenzamide

Safety and Handling

Recommended Storage : Store at +4°C