Please login to your online account to display your discounted pricing

Alfa Aesar™ o-Xylylenebis(triphenylphosphonium bromide), 98+%

$57.57 - $57.57

Chemical Identifiers

Chemical Name or Material o-Xylylenebis(triphenylphosphonium bromide)
CAS 1519-46-6
Molecular Formula C44H38Br2P2
Formula Weight 788.55
MDL Number MFCD00044843
Beilstein 4121635
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0310309

  SDS
alfa aesar™
L0310309
10g Each for $57.57
Description & Specifications

Specifications

Formula Weight 788.55
MDL Number MFCD00044843
Molecular Formula C44H38Br2P2
Chemical Name or Material o-Xylylenebis(triphenylphosphonium bromide)
Assay ≥98%
CAS 1519-46-6
Quantity 10g
Health Hazard 1 Warning
Odor Odorless
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 4121635
Melting Point >300°C
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.