Please login to your online account to display your discounted pricing

Alfa Aesar™ o-Xylylene dichloride, 98%

Chemical Identifiers

Chemical Name or Material o-Xylylene dichloride
CAS 612-12-4
Molecular Formula C8H8Cl2
Formula Weight 175.06
MDL Number MFCD00000903
UN Number UN2928
Beilstein 2043675
Synonym 1,2-Bis(chloromethyl)benzene; alpha,alpha'-Dichloro-o-xylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1250018

  SDS
alfa aesar™
A1250018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1250030

  SDS
alfa aesar™
A1250030
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.06
Chemical Name or Material o-Xylylene dichloride
TSCA TSCA
Assay 98%
Molecular Formula C8H8Cl2
MDL Number MFCD00000903
CAS 612-12-4
UN Number UN2928
Beilstein 2043675
Synonym 1,2-Bis(chloromethyl)benzene; alpha,alpha'-Dichloro-o-xylene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H330-H314-H318-H302
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280-P273-P305+P351+P338-P309-P310-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 246°C
Melting Point 53° to 57°C
Flash Point 107°C (224°F)
Odor Pungent
Quantity 50g
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
RTECS Number ZE4085000
Safety and Handling

Danger

GHS P Statement
P280-P273-P305+P351+P338-P309-P310-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H330-H314-H318-H302
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.