missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Xylene, Spectrophotometric Grade, 96% min, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 032471K7

 View more versions of this product

Catalog No. AA32471K7



Description

Description

o-Xylene is used as a solvent for paints and adhesives, and to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

o-Xylene
95-47-6
0.877 g/mL
4 L
Liquid
C8H10
MFCD00008519
1815558
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
CTQNGGLPUBDAKN-UHFFFAOYSA-N
1,2-xylene
7237
106.17
Spectrophotometric
-25°C
Colorless
143°C to 145°C
32°C (89°F)
1.505
(CH3)2C6H4
UN1307
14,10081
Sparingly soluble in water. (0.2g/L)
CC1=CC=CC=C1C
106.17
CHEBI:28063
≤96%

Chemical Identifiers

95-47-6
106.17
CTQNGGLPUBDAKN-UHFFFAOYSA-N
7237
1,2-xylene
C8H10
MFCD00008519
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
CHEBI:28063
CC1=CC=CC=C1C
Safety and Handling

Safety and Handling

GHS H Statement
H226-H312-H332-H315
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c

H226-H304-H312+H332-H315-H319-H335-H336-H373

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: XYLENES

EINECSNumber : 202-422-2

RTECSNumber : ZE2450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only