missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ o-Xylene, Spectrophotometric Grade, 96% min

Manufacturer:  Alfa Aesar™ 32471K2

 View more versions of this product

Catalog No. AA32471K2



Specifications

Specifications

o-Xylene
Spectrophotometric grade
C8H10
MFCD00008519
1815558
CTQNGGLPUBDAKN-UHFFFAOYSA-N
1,2-xylene
7237
106.17
0.877g/mL
32° (89°F)
Colorless
1L
95-47-6
≤96%
(CH3)2C6H4
UN1307
14,10081
CC1=CC=CC=C1C
106.168
CHEBI:28063
Liquid
1.5050
143°C to 145°C
-25°C

Chemical Identifiers

95-47-6
106.168
CTQNGGLPUBDAKN-UHFFFAOYSA-N
CHEBI:28063
CC1=CC=CC=C1C
C8H10
MFCD00008519
7237
1,2-xylene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H312-H332-H315
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: XYLENES

RTECSNumber : ZE2450000

TSCA : TSCA

SDS Documents