Please login to your online account to display your discounted pricing

o-Xylene-d10, for NMR, 99+ atom % D, ACROS Organics™

$124.30 - $124.30

Chemical Identifiers

Chemical Name or Material o-Xylene-d10
Name Note For NMR
CAS 56004-61-6
Purity Grade For NMR
Molecular Formula C8D10
Linear Formula C6D4(CD3)2
Formula Weight 116.25g/mol
MDL Number MFCD00037698
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC174860050

  SDS
thermo fisher scientific
174860050
Glass bottle 5g Each for $124.30
Description & Specifications

Specifications

Deuterium (D) 99.0% min.
Packaging Glass bottle
Density 0.9530g/mL
Quantity 5g
Color Clear colorless
Chemical Name or Material o-Xylene-d10
Name Note For NMR
CAS 56004-61-6
Purity Grade For NMR
Physical Form Clear colorless liquid
Molecular Formula C8D10
Linear Formula C6D4(CD3)2
Formula Weight 116.25g/mol
MDL Number MFCD00037698
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

For NMR

Safety and Handling

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
Flammable liquid and vapor.