Please login to your online account to display your discounted pricing

Alfa Aesar™ o-Xylene-d{10}, 98+% (Isotopic)

Chemical Identifiers

Chemical Name or Material o-Xylene-d{10}
CAS 56004-61-6
Name Note Isotopic
Formula Weight 116.25
MDL Number MFCD00037698
UN Number UN1307
Merck Index 14,10081
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3647206

  SDS
alfa aesar™
3647206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material o-Xylene-d{10}
Name Note Isotopic
CAS 56004-61-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H312-H332-H315
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: XYLENES
Sensitivity Moisture Sensitive
Density 0.953g/mL
Boiling Point 143° to 145°C
Melting Point −25°C
Physical Form Liquid
Assay ≥98%
Quantity 5g
Color Colorless
Formula Weight 116.25
MDL Number MFCD00037698
UN Number UN1307
Merck Index 14,10081
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H312-H332-H315
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.