missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Xylene, 96% min., HPLC Grade, Alfa Aesar™

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.04%

Manufacturer:  Alfa Aesar 22902M1

 View more versions of this product

Catalog No. AA22902M1



Specifications

Specifications

o-Xylene
Colorless liquid
C8H10
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
CC1=CC=CC=C1C
106.168
CHEBI:28063
Liquid
0.877g/cm3
Characteristic
Colorless
95-47-6
96% min.
MFCD00008519
CTQNGGLPUBDAKN-UHFFFAOYSA-N
1,2-xylene
7237
106.17
HPLC
465°C (869°F)
6.7 hPa (5mm Hg) at 20°C
4 x 1L

Chemical Identifiers

95-47-6
106.168
CTQNGGLPUBDAKN-UHFFFAOYSA-N
7237
1,2-xylene
C8H10
MFCD00008519
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
CHEBI:28063
CC1=CC=CC=C1C
Safety and Handling

Safety and Handling

H226-H312-H332-H315: Flammable liquid and vapour. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation.

DOTInformation : Hazard Class: 3, Packing Group: II

EINECSNumber : 202-422-2

SDS Documents