missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-(Trimethylsilyl)hydroxylamine 95.0+%, TCI America™
Supplier: TCI America T16695G
Specifications
| O-(Trimethylsilyl)hydroxylamine | |
| Colorless | |
| C3H11NOSi | |
| 5 g | |
| o-trimethylsilyl hydroxylamine, hydroxylamine, o-trimethylsilyl, aminoxytrimethylsilane, o-tms-hydroxylamine, tmsonh2, acmc-1clek, aminooxy trimethylsilane, 0-trimethylsilylhydroxylamine, o-trmethylslyl hydroxylamne | |
| C[Si](C)(C)ON | |
| 105.21 | |
| 105.21 | |
| Liquid |
| 22737-36-6 | |
| 105°C | |
| MFCD00010872 | |
| 2733 | |
| AEKHNNJSMVVESS-UHFFFAOYSA-N | |
| O-(trimethylsilyl)hydroxylamine | |
| 2734666 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 22737-36-6 | |
| 105.21 | |
| AEKHNNJSMVVESS-UHFFFAOYSA-N | |
| 2734666 | |
| C[Si](C)(C)ON |
| C3H11NOSi | |
| MFCD00010872 | |
| o-trimethylsilyl hydroxylamine, hydroxylamine, o-trimethylsilyl, aminoxytrimethylsilane, o-tms-hydroxylamine, tmsonh2, acmc-1clek, aminooxy trimethylsilane, 0-trimethylsilylhydroxylamine, o-trmethylslyl hydroxylamne | |
| O-(trimethylsilyl)hydroxylamine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator