missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-(Trimethylsilyl)hydroxylamine 95.0+%, TCI America™
$367.22 - $1243.32
Chemical Identifiers
| CAS | 22737-36-6 |
|---|---|
| Molecular Formula | C3H11NOSi |
| Molecular Weight (g/mol) | 105.21 |
| MDL Number | MFCD00010872 |
| InChI Key | AEKHNNJSMVVESS-UHFFFAOYSA-N |
| Synonym | o-trimethylsilyl hydroxylamine, hydroxylamine, o-trimethylsilyl, aminoxytrimethylsilane, o-tms-hydroxylamine, tmsonh2, acmc-1clek, aminooxy trimethylsilane, 0-trimethylsilylhydroxylamine, o-trmethylslyl hydroxylamne |
| PubChem CID | 2734666 |
| IUPAC Name | O-(trimethylsilyl)hydroxylamine |
| SMILES | C[Si](C)(C)ON |
Chemical Identifiers
| 22737-36-6 | |
| 105.21 | |
| AEKHNNJSMVVESS-UHFFFAOYSA-N | |
| 2734666 | |
| C[Si](C)(C)ON |
| C3H11NOSi | |
| MFCD00010872 | |
| o-trimethylsilyl hydroxylamine, hydroxylamine, o-trimethylsilyl, aminoxytrimethylsilane, o-tms-hydroxylamine, tmsonh2, acmc-1clek, aminooxy trimethylsilane, 0-trimethylsilylhydroxylamine, o-trmethylslyl hydroxylamne | |
| O-(trimethylsilyl)hydroxylamine |
Specifications
| 22737-36-6 | |
| 105°C | |
| MFCD00010872 | |
| 2733 | |
| AEKHNNJSMVVESS-UHFFFAOYSA-N | |
| O-(trimethylsilyl)hydroxylamine | |
| 2734666 | |
| ≥95.0% (GC) | |
| O-(Trimethylsilyl)hydroxylamine |
| Colorless | |
| C3H11NOSi | |
| 5 g | |
| o-trimethylsilyl hydroxylamine, hydroxylamine, o-trimethylsilyl, aminoxytrimethylsilane, o-tms-hydroxylamine, tmsonh2, acmc-1clek, aminooxy trimethylsilane, 0-trimethylsilylhydroxylamine, o-trmethylslyl hydroxylamne | |
| C[Si](C)(C)ON | |
| 105.21 | |
| 105.21 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator