missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

O-(Trimethylsilyl)hydroxylamine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC367950010

 View more versions of this product

Catalog No. AC367950010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

O-(Trimethylsilyl)hydroxylamine
Clear colorless
97%
96.0
Authentic
1.4040 to 1.4060 (20°C, 589nm)
≥96.0%
C3H11NOSi
MFCD00010872
0.86g/mL
1g
22737-36-6
100.0
Glass bottle
Liquid
97%
105.21
Aminoxytrimethylsilane

Chemical Identifiers

22737-36-6
100.0
C3H11NOSi
MFCD00010872
O-(Trimethylsilyl)hydroxylamine, 97%
96.0
97%
105.21
Aminoxytrimethylsilane
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger