Please login to your online account to display your discounted pricing

Alfa Aesar™ o-Tolylurea, 98%

$38.18 - $280.15

Chemical Identifiers

Chemical Name or Material o-Tolylurea
CAS 614-77-7
Molecular Formula C8H10N2O
Formula Weight 150.18
MDL Number MFCD00025407
Beilstein 2690754
Synonym 2-Methylphenylurea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2308606

 
alfa aesar™
B2308606
5g Each for $38.18

AAB2308622

 
alfa aesar™
B2308622
100g Each for $280.15
Description & Specifications

Specifications

Formula Weight 150.18
Chemical Name or Material o-Tolylurea
Assay 98%
Molecular Formula C8H10N2O
MDL Number MFCD00025407
CAS 614-77-7
Beilstein 2690754
Synonym 2-Methylphenylurea
Melting Point 198° to 200°C
Quantity 5g
RTECS Number YU4100000