missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Tolylmagnesium chloride, 1.4M (22 wt.%) solution in THF/toluene, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC427488000

 View more versions of this product

Catalog No. AC427488000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

150.89
AcroSeal™ Glass Bottle
109-99-9,33872-80-9,108-88-3
CH3C6H4MgCl
C7H7ClMg
brown solution
800mL
0.9600g/mL
o-Tolylmagnesium chloride
1.4M (22 wt.%) solution in THF/toluene
MFCD00010355

Chemical Identifiers

33872-80-9,109-99-9,108-88-3
CH3C6H4MgCl
MFCD00010355
C7H7ClMg
150.89
o-Tolylmagnesium chloride, 1.4M (22 wt.%) solution in THF/toluene

Safety and Handling

SDS & Safety Documents

GHS H Statement
Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. May cause respiratory irritation. May cause damage to organs through prolonged or repeated exposure. Suspected of damaging the unborn child. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Suspected of causing cancer.

GHS P Statement
Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Do not breathe dust/fume/gas/mist/vapors/spray. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a dry place. Store in a closed container.

Danger