Please login to your online account to display your discounted pricing

o-Tolylmagnesium chloride, 1.4M (22 wt.%) solution in THF/toluene, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC427481000

 View more versions of this product

Catalog No. AC427481000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 150.89
Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 0.9600g/mL
Chemical Name or Material o-Tolylmagnesium chloride
Name Note 1.4M (22 wt.%) solution in THF/toluene
Appearance of Solution brown solution
CAS 108-88-3
Molecular Formula C7H7ClMg
Linear Formula CH3C6H4MgCl
MDL Number MFCD00010355
CAS 33872-80-9
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. May cause respiratory irritation. May cause damage to organs through prolonged or repeated exposure. Suspected of
CAS 109-99-9
Health Hazard 3 GHS P Statement Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

Specifications

Chemical Name or Material o-Tolylmagnesium chloride
Name Note 1.4M (22 wt.%) solution in THF/toluene
CAS 33872-80-9
CAS 109-99-9
CAS 108-88-3
Molecular Formula C7H7ClMg
Linear Formula CH3C6H4MgCl
Formula Weight 150.89
MDL Number MFCD00010355
Accessories

Danger

GHS P Statement Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

GHS P Statement Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. May cause respiratory irritation. May cause damage to organs through prolonged or repeated exposure. Suspected of

SDS & Safety Documents