Please login to your online account to display your discounted pricing

o-Tolylhydrazine Hydrochloride 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material o-Tolylhydrazine hydrochloride
Name Note 98%
CAS 635-26-7
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C7H10N2·HCl
Linear Formula CH3C6H4NHNH2·HCl
Formula Weight 158.63g/mol
MDL Number MFCD00012925
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC139140100

  SDS
thermo fisher scientific
139140100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color Beige
Quantity 10g
Chemical Name or Material o-Tolylhydrazine hydrochloride
Name Note 98%
CAS 635-26-7
Physical Form Beige crystalline powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C7H10N2·HCl
Linear Formula CH3C6H4NHNH2·HCl
Formula Weight 158.63g/mol
MDL Number MFCD00012925
Beilstein 15, 496
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 211-232-9
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.