Please login to your online account to display your discounted pricing

Alfa Aesar™ o-Tolylhydrazine hydrochloride, 98%

Chemical Identifiers

Chemical Name or Material o-Tolylhydrazine hydrochloride
CAS 635-26-7
Molecular Formula C7H10N2·HCl
Formula Weight 158.63
MDL Number MFCD00012925
UN Number UN2811
Beilstein 3655929
Synonym 2-Methylphenylhydrazine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1576714

  SDS
alfa aesar™
A1576714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1576722

  SDS
alfa aesar™
A1576722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 158.63
Synonym 2-Methylphenylhydrazine hydrochloride
UN Number UN2811
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈190°C (dec.)
Chemical Name or Material o-Tolylhydrazine hydrochloride
CAS 635-26-7
Health Hazard 1 Warning
Quantity 25g
Molecular Formula C7H10N2·HCl
Beilstein 3655929
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
MDL Number MFCD00012925
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.