missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ o-Tolylacetyl chloride, 98%

Manufacturer:  Alfa Aesar™ H37418MD

 View more versions of this product

Catalog No. AAH37418MD



Specifications

Specifications

o-Tolylacetyl chloride
84°C (1.5mmHg)
250mg
C9H9ClO
UN3265
FKMIBYMTHOJWCY-UHFFFAOYSA-N
2-(2-methylphenyl)acetyl chloride
2760632
Liquid
Moisture Sensitive
Pink
10166-09-3
MFCD03424714
o-tolyl-acetyl chloride, o-tolylacetyl chloride, 2-2-methylphenyl acetyl chloride, o-methylphenylacetyl chloride, 2-methylphenylacetyl chloride, 2-methylbenzeneacetyl chloride, 2-methylphenyl acetyl chloride, 2-methylphenylacetic acid chloride
CC1=CC=CC=C1CC(=O)Cl
168.62
168.62
98%

Chemical Identifiers

10166-09-3
168.62
FKMIBYMTHOJWCY-UHFFFAOYSA-N
2760632
CC1=CC=CC=C1CC(=O)Cl
C9H9ClO
MFCD03424714
o-tolyl-acetyl chloride, o-tolylacetyl chloride, 2-2-methylphenyl acetyl chloride, o-methylphenylacetyl chloride, 2-methylphenylacetyl chloride, 2-methylbenzeneacetyl chloride, 2-methylphenyl acetyl chloride, 2-methylphenylacetic acid chloride
2-(2-methylphenyl)acetyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

SDS Documents