Learn More
o-Tolyl isothiocyanate, 99%, Thermo Scientific Chemicals
$56.75 - $56.75
Chemical Identifiers
CAS | 614-69-7 |
---|---|
MDL Number | MFCD00004802 |
InChI Key | JYKYYPPZLPVIBY-UHFFFAOYSA-N |
Synonym | 2-methylphenyl isothiocyanate, o-tolyl isothiocyanate, 2-tolyl isothiocyanate, o-tolylisothiocyanate, benzene, 1-isothiocyanato-2-methyl, 1-isothiocyanato-2-methyl-benzene, 2-methylphenylisothiocyanate, isothiocyanic acid o-tolyl ester, isothiocyanic acid, o-tolyl ester, 2-methylbenzenisothiocyanate |
PubChem CID | 69195 |
IUPAC Name | 1-isothiocyanato-2-methylbenzene |
SMILES | CC1=CC=CC=C1N=C=S |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC292650050
|
Thermo Scientific Chemicals
292650050 |
5 g |
Each for $56.75
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
614-69-7 | |
JYKYYPPZLPVIBY-UHFFFAOYSA-N | |
69195 | |
CC1=CC=CC=C1N=C=S |
MFCD00004802 | |
2-methylphenyl isothiocyanate, o-tolyl isothiocyanate, 2-tolyl isothiocyanate, o-tolylisothiocyanate, benzene, 1-isothiocyanato-2-methyl, 1-isothiocyanato-2-methyl-benzene, 2-methylphenylisothiocyanate, isothiocyanic acid o-tolyl ester, isothiocyanic acid, o-tolyl ester, 2-methylbenzenisothiocyanate | |
1-isothiocyanato-2-methylbenzene |
Specifications
614-69-7 | |
1.1100g/mL | |
107°C | |
99% | |
C8H7NS | |
CH3C6H4NCS | |
5 g | |
2-methylphenyl isothiocyanate, o-tolyl isothiocyanate, 2-tolyl isothiocyanate, o-tolylisothiocyanate, benzene, 1-isothiocyanato-2-methyl, 1-isothiocyanato-2-methyl-benzene, 2-methylphenylisothiocyanate, isothiocyanic acid o-tolyl ester, isothiocyanic acid, o-tolyl ester, 2-methylbenzenisothiocyanate | |
JYKYYPPZLPVIBY-UHFFFAOYSA-N | |
1-isothiocyanato-2-methylbenzene | |
69195 | |
99% | |
o-Tolyl isothiocyanate |
Yellow | |
239°C | |
Authentic | |
Glass bottle | |
1.6351 to 1.6371 | |
MFCD00004802 | |
1.11 | |
Solubility in water: may decompose | |
CC1=CC=CC=C1N=C=S | |
149.22 | |
149.22 | |
Liquid |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
Harmful in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quali
GHS Signal Word: Danger
EINECSNumber : 210-390-6
RUO – Research Use Only