missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Tolyl isothiocyanate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC292650050

 View more versions of this product

Catalog No. AC292650050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

o-Tolyl isothiocyanate
107°C
Glass bottle
1.11
Yellow
614-69-7
C8H7NS
MFCD00004802
Solubility in water: may decompose
CC1=CC=CC=C1N=C=S
149.22
149.22
99%
1.1100g/mL
Authentic
1.6351 to 1.6371
239°C
5g
99%
CH3C6H4NCS
1-isothiocyanato-2-methyl-benzene, 2-methylbenzenisothiocyanate, 2-methylphenyl isothiocyanate, 2-methylphenylisothiocyanate, 2-tolyl isothiocyanate, benzene, 1-isothiocyanato-2-methyl, isothiocyanic acid o-tolyl ester, isothiocyanic acid, o-tolyl ester, o-tolyl isothiocyanate, o-tolylisothiocyanate
JYKYYPPZLPVIBY-UHFFFAOYSA-N
1-isothiocyanato-2-methylbenzene
69195
Liquid

Chemical Identifiers

614-69-7
149.22
JYKYYPPZLPVIBY-UHFFFAOYSA-N
69195
CC1=CC=CC=C1N=C=S
C8H7NS
MFCD00004802
1-isothiocyanato-2-methyl-benzene, 2-methylbenzenisothiocyanate, 2-methylphenyl isothiocyanate, 2-methylphenylisothiocyanate, 2-tolyl isothiocyanate, benzene, 1-isothiocyanato-2-methyl, isothiocyanic acid o-tolyl ester, isothiocyanic acid, o-tolyl ester, o-tolyl isothiocyanate, o-tolylisothiocyanate
1-isothiocyanato-2-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement:
May cause an allergic skin reaction.
Harmful in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-390-6

SDS Documents