missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Tolyl isocyanate, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 292220250

 View more versions of this product

Catalog No. AC292220250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

o-Tolyl isocyanate
92°C
Glass bottle
1.074
Colorless to Yellow
99+%
99% min. (GC)
CH3C6H4NCO
12, 812
VAYMIYBJLRRIFR-UHFFFAOYSA-N
1-isocyanato-2-methylbenzene
69194
133.15
99+%
1.0740g/mL
Authentic
1.5335 to 1.5355
185°C
25g
614-68-6
C8H7NO
MFCD00002009
2-methylphenyl isocyanate, 2-methylphenylisocyanate, 2-tolyl isocyanate, benzene, 1-isocyanato-2-methyl, isocyanic acid, o-tolyl ester, o-methylphenyl isocyanate, o-toluene isocyanate, o-tolyl isocyanate, o-tolylisocyanate, tolyl isocyanate
CC1=CC=CC=C1N=C=O
133.15
CHEBI:60099
Liquid

Chemical Identifiers

614-68-6
133.15
VAYMIYBJLRRIFR-UHFFFAOYSA-N
69194
1-isocyanato-2-methylbenzene
C8H7NO
MFCD00002009
2-methylphenyl isocyanate, 2-methylphenylisocyanate, 2-tolyl isocyanate, benzene, 1-isocyanato-2-methyl, isocyanic acid, o-tolyl ester, o-methylphenyl isocyanate, o-toluene isocyanate, o-tolyl isocyanate, o-tolylisocyanate, tolyl isocyanate
CHEBI:60099
CC1=CC=CC=C1N=C=O
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Danger

EINECSNumber : 210-389-0

RTECSNumber : DA3678333

TSCA : TSCA

Recommended Storage : Product may develop some turbidity or precipitate

SDS Documents