Please login to your online account to display your discounted pricing

Alfa Aesar™ o-Tolyl benzoate, 99%

$46.26 - $131.23

Chemical Identifiers

Chemical Name or Material o-Tolyl benzoate
CAS 617-02-7
Molecular Formula C14H12O2
Formula Weight 212.25
MDL Number MFCD00041330
Beilstein 1873222
Synonym Benzoic acid o-tolyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1183414

  SDS
alfa aesar™
L1183414
25g Each for $46.26

AAL1183422

  SDS
alfa aesar™
L1183422
100g Each for $131.23
Description & Specifications

Specifications

Formula Weight 212.25
Chemical Name or Material o-Tolyl benzoate
MDL Number MFCD00041330
CAS 617-02-7
Assay 99%
Molecular Formula C14H12O2
Beilstein 1873222
Synonym Benzoic acid o-tolyl ester
Boiling Point 170° to 172°C (11mmHg)
Quantity 25g