missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ o-Tolyl benzoate, 99%

Manufacturer:  Alfa Aesar™ L1183422

 View more versions of this product

Catalog No. AAL1183422



Specifications

Specifications

o-Tolyl benzoate
100g
C14H12O2
1873222
YPMKPCAILYDVBN-UHFFFAOYSA-N
(2-methylphenyl) benzoate
69231
99%
170°C to 172°C (11mmHg)
617-02-7
MFCD00041330
o-tolyl benzoate, benzoic acid, o-tolyl ester, o-tolylbenzoate, benzoic acid, 2-methylphenyl ester, toluene benzoate, acmc-1az7o, 2-methylphenyl benzoate, benzoic acid o-tolyl ester, ortho-tolyl benzoate
CC1=CC=CC=C1OC(=O)C2=CC=CC=C2
212.248
212.25

Chemical Identifiers

617-02-7
212.248
YPMKPCAILYDVBN-UHFFFAOYSA-N
69231
CC1=CC=CC=C1OC(=O)C2=CC=CC=C2
C14H12O2
MFCD00041330
o-tolyl benzoate, benzoic acid, o-tolyl ester, o-tolylbenzoate, benzoic acid, 2-methylphenyl ester, toluene benzoate, acmc-1az7o, 2-methylphenyl benzoate, benzoic acid o-tolyl ester, ortho-tolyl benzoate
(2-methylphenyl) benzoate
Documents