missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Tolyl 3,5-Xylyl Ether, TCI America™

Manufacturer:  TCI America T221325G

 View more versions of this product

Catalog No. T221325G



Specifications

Specifications

o-Tolyl 3,5-Xylyl Ether
25g
C15H16O
YCAZRHVXMXMPEY-UHFFFAOYSA-N
1,3-dimethyl-5-(2-methylphenoxy)benzene
15324946
Clear Liquid at 20°C
Colorless
196604-20-3
2,3′,5′-Trimethyldiphenyl Ether
CC1=CC=CC=C1OC2=CC(=CC(=C2)C)C
212.292
212.29
≥97.0% (GC)

Chemical Identifiers

196604-20-3
212.292
2,3′,5′-Trimethyldiphenyl Ether
1,3-dimethyl-5-(2-methylphenoxy)benzene
C15H16O
YCAZRHVXMXMPEY-UHFFFAOYSA-N
15324946
CC1=CC=CC=C1OC2=CC(=CC(=C2)C)C
Safety and Handling

Safety and Handling

TSCA : No

Documents