missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Tolunitrile 98%, ACROS Organics™

Chemical Identifiers

CAS 529-19-1
Assay Percent Range 98%
Molecular Formula C8H7N
Linear Formula CH3C6H4CN
Formula Weight 117.15
MDL Number MFCD00001792
Synonym 2-Methylbenzenecarbonitrile
Chemical Name or Material o-Tolunitrile, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC152860010 SDS Thermo Fisher Scientific
AC152860010
Glass bottle 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC152860250 SDS Fisher Scientific Acros
152860250
Glass bottle 25mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC152862500 SDS Fisher Scientific Acros
152862500
Glass bottle 250mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

529-19-1
C8H7N
117.15
2-Methylbenzenecarbonitrile
≥97.5%
98%
CH3C6H4CN
MFCD00001792
o-Tolunitrile, 98%

Specifications

529-19-1
Liquid
C8H7N
Glass bottle
MFCD00001792
1.5269 to 1.5289 (20°C, 589nm)
2-Methylbenzenecarbonitrile
Clear colorless to slightly yellow
o-Tolunitrile, 98%
Authentic
98%
CH3C6H4CN
117.15
09, 466
15, 9697
0.99g/mL
1L
≥97.5%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 208-451-7

RTECSNumber : XV0600000

TSCA : TSCA