Please login to your online account to display your discounted pricing

Alfa Aesar™ o-Tolunitrile, 98%

$68.86 - $68.86

Chemical Identifiers

Chemical Name or Material o-Tolunitrile
CAS 529-19-1
Molecular Formula C8H7N
Formula Weight 117.15
MDL Number MFCD00001792
Beilstein 1857417
Merck Index 14,9537
Synonym 2-Methylbenzonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2223130

  SDS
alfa aesar™
B2223130
250g Each for $68.86
Description & Specifications

Specifications

Formula Weight 117.15
Odor Almond-like
Flash Point 84°C (183°F)
Melting Point −14°C
Assay 98%
Molecular Formula C8H7N
Health Hazard 3 GHS P Statement
P261-P280g-P273-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Boiling Point 205° to 206°C
MDL Number MFCD00001792
Chemical Name or Material o-Tolunitrile
Synonym 2-Methylbenzonitrile
RTECS Number XV0600000
Quantity 250g
Beilstein 1857417
CAS 529-19-1
Merck Index 14,9537
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
TSCA TSCA
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280g-P273-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.