missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Toluenethiol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC259240250

 View more versions of this product

Catalog No. AC259240250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

o-Toluenethiol
63°C
Glass bottle
1.054
Colorless to Yellow
25g
137-06-4
C7H8S
06,37
2-methylthiophenol, 2-thiocresol, 2-toluenethiol, benzenethiol, 2-methyl, o-mercaptotoluene, o-methylbenzenethiol, o-methylthiophenol, o-thiocresol, o-toluenethiol, o-tolyl mercaptan
LXUNZSDDXMPKLP-UHFFFAOYSA-N
2-methylbenzenethiol
8712
Liquid
1.0540g/mL
Authentic
1.5790 to 1.5810
194.0°C
14.0°C
97%
96% min. (GC)
CH3C6H4SH
15,9478
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
CC1=CC=CC=C1S
124.21
124.21
97%

Chemical Identifiers

137-06-4
124.21
2-methylthiophenol, 2-thiocresol, 2-toluenethiol, benzenethiol, 2-methyl, o-mercaptotoluene, o-methylbenzenethiol, o-methylthiophenol, o-thiocresol, o-toluenethiol, o-tolyl mercaptan
2-methylbenzenethiol
C7H8S
LXUNZSDDXMPKLP-UHFFFAOYSA-N
8712
CC1=CC=CC=C1S
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED: rinse mouth.
Do

GHS Signal Word: Danger

EINECSNumber : 205-276-8

RTECSNumber : XT8750000

TSCA : TSCA

SDS Documents