Learn More
o-Tolualdehyde, 98%, stabilized, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 152431000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
o-Tolualdehyde | |
529-20-4,123-31-9 | |
199°C to 200°C | |
Authentic | |
Glass bottle | |
1.5460 to 1.548 | |
MFCD00003338 | |
07, 295 | |
15, 9686 | |
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol and ether | |
CC1=CC=CC=C1C=O | |
120.15 | |
10722 | |
120.15 |
98% | |
1.0390g/mL | |
67°C | |
97.5% min. (GC) | |
C8H8O | |
CH3C6H4CHO | |
100g | |
1.039 | |
o-tolualdehyde, 2-tolualdehyde, 2-formyltoluene, o-toluylaldehyde, o-toluic aldehyde, o-tolylaldehyde, o-methylbenzaldehyde, benzaldehyde, 2-methyl, 2-methyl-benzaldehyde, o-methylbenazldehyde | |
BTFQKIATRPGRBS-UHFFFAOYSA-N | |
2-methylbenzaldehyde | |
0.1% hydroquinone | |
CHEBI:27434 | |
98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May produce an allergic reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen
GHS Signal Word: Warning
EINECSNumber : 208-452-2
RUO â Research Use Only