missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ o-Tolualdehyde, 98%, stab. with 0.1% hydroquinone

Manufacturer:  Alfa Aesar™ A1017222

 View more versions of this product

Catalog No. AAA1017222



Description

Description

Applications
o-tolualdehyde was used in determination of alkenal-2,4-dinitrophenylhydrazones by HPLC by addition of phosphoric acid. It produces to give a single diastereomer of the tetrahydronaphthalene derivative under UV irradiation.

Solubility
Soluble in carbon tetrachloride, ethanol, ethyl ether, benzene. Not miscible in water.

Notes
Air Sensitive. Incompatible with air and oxidizing agents. Store under dry inert gas.
Specifications

Specifications

o-Tolualdehyde
Almond-like
-35°C
Stabilized with 0.1% hydroquinone
C8H8O
605841
o-tolualdehyde, 2-tolualdehyde, 2-formyltoluene, o-toluylaldehyde, o-toluic aldehyde, o-tolylaldehyde, o-methylbenzaldehyde, benzaldehyde, 2-methyl, 2-methyl-benzaldehyde, o-methylbenazldehyde
CC1=CC=CC=C1C=O
120.151
CHEBI:27434
98%
67°C (152°F)
199°C to 200°C
100g
529-20-4
MFCD00003338
14,9527
BTFQKIATRPGRBS-UHFFFAOYSA-N
2-methylbenzaldehyde
10722
120.15

Chemical Identifiers

529-20-4
120.151
BTFQKIATRPGRBS-UHFFFAOYSA-N
10722
2-methylbenzaldehyde
C8H8O
MFCD00003338
o-tolualdehyde, 2-tolualdehyde, 2-formyltoluene, o-toluylaldehyde, o-toluic aldehyde, o-tolylaldehyde, o-methylbenzaldehyde, benzaldehyde, 2-methyl, 2-methyl-benzaldehyde, o-methylbenazldehyde
CHEBI:27434
CC1=CC=CC=C1C=O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents