missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Tolualdehyde 2,4-Dinitrophenylhydrazone, TCI America™

Manufacturer:  TCI America T144710G

 View more versions of this product

Catalog No. T144710G



Specifications

Specifications

o-Tolualdehyde 2,4-Dinitrophenylhydrazone
196°C
1773-44-0
MFCD00191584
2′-Methylbenzaldehyde 2,4-Dinitrophenylhydrazone
CC1=CC=CC=C1C=NNC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]
300.274
300.27
≥98.0% (HPLC,T)
Undesignated
10g
C14H12N4O4
1325
QUIFNJPMGQCNMH-OQLLNIDSSA-N
N-[(E)-(2-methylphenyl)methylideneamino]-2,4-dinitroaniline
9555686
Crystal-Powder at 20°C

Chemical Identifiers

1773-44-0
300.274
QUIFNJPMGQCNMH-OQLLNIDSSA-N
9555686
CC1=CC=CC=C1C=NNC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]
C14H12N4O4
MFCD00191584
2′-Methylbenzaldehyde 2,4-Dinitrophenylhydrazone
N-[(E)-(2-methylphenyl)methylideneamino]-2,4-dinitroaniline
Safety and Handling

Safety and Handling

TSCA : No

Documents