missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Tolidine 98.0+%, TCI America™

$75.05 - $673.89

Chemical Identifiers

CAS 119-93-7
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.296
MDL Number MFCD00014773
InChI Key NUIURNJTPRWVAP-UHFFFAOYSA-N
Synonym 2-tolidine, 3,3'-dimethylbenzidine, 3,3'-dimethylbiphenyl-4,4'-diamine, 3,3'-tolidine, 4,4'-bi-o-toluidine, bianisidine, diaminoditolyl, diaminotolyl, o-tolidine, orthotolidine
PubChem CID 8413
ChEBI CHEBI:34320
IUPAC Name 4-(4-amino-3-methylphenyl)-2-methylaniline
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T191325G
View Documents
Tci America
T191325G
25g
Each for $75.05
 
T1913500G
View Documents
Tci America
T1913500G
500g
Each for $673.89
 
Description

Description

Specifications

Chemical Identifiers

119-93-7
212.296
NUIURNJTPRWVAP-UHFFFAOYSA-N
8413
4-(4-amino-3-methylphenyl)-2-methylaniline
C14H16N2
MFCD00014773
2-tolidine, 3,3'-dimethylbenzidine, 3,3'-dimethylbiphenyl-4,4'-diamine, 3,3'-tolidine, 4,4'-bi-o-toluidine, bianisidine, diaminoditolyl, diaminotolyl, o-tolidine, orthotolidine
CHEBI:34320
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N

Specifications

300°C
130°C
119-93-7
MFCD00014773
2-tolidine, 3,3'-dimethylbenzidine, 3,3'-dimethylbiphenyl-4,4'-diamine, 3,3'-tolidine, 4,4'-bi-o-toluidine, bianisidine, diaminoditolyl, diaminotolyl, o-tolidine, orthotolidine
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N
212.296
CHEBI:34320
Crystal-Powder at 20°C
o-Tolidine
Undesignated
25g
C14H16N2
3077
NUIURNJTPRWVAP-UHFFFAOYSA-N
4-(4-amino-3-methylphenyl)-2-methylaniline
8413
212.30
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0882

RTECSNumber : DD1225000

TSCA : Yes

SDS