Please login to your online account to display your discounted pricing

o-Tolidine Dihydrochloride Solution (0.1% w/v), Fisher Chemical


For determination of chlorine in water

Quantity 500mL
Packaging Glass Bottle

Manufacturer: fisher scientific company  SO14500

 View more versions of this product

Catalog No. SO14-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Melting Point −10°C
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
pH 1
Alpha Vector Tolidine Dihydrochloride Solution
Density 1.03g/cm³
Color Clear
Packaging Glass Bottle
Sensitivity Pass Test
MDL Number MFCD00012960
ChemAlert Storage Symbol White
Concentration or Composition (by Analyte or Components) 0.1% w/v
CAS 7732-18-5
Chemical Name or Material o-Tolidine Dihydrochloride Solution
Name Note 0.1% (w/v)
CAS 7647-01-0
CAS 612-82-8

Specifications

Chemical Name or Material o-Tolidine Dihydrochloride Solution
Name Note 0.1% (w/v)
CAS 7647-01-0
CAS 612-82-8
CAS 7732-18-5
MDL Number MFCD00012960
Accessories
 

SDS & Safety Documents