Please login to your online account to display your discounted pricing

o-Tolidine Dihydrochloride Solution (0.1% w/v), Fisher Chemical

For determination of chlorine in water

$147.85 - $195.31

Chemical Identifiers

Chemical Name or Material o-Tolidine Dihydrochloride Solution
Name Note 0.1% (w/v)
CAS 7647-01-0
CAS 612-82-8
CAS 7732-18-5
MDL Number MFCD00012960
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SO14-1

  SDS
fisher scientific company
SO141
Glass Bottle 1L Each for $195.31
Case of 6 Each for $1,171.86

SO14-500

  SDS
fisher scientific company
SO14500
Glass Bottle 500mL Each for $147.85
Case of 6 Each for $887.10
Description & Specifications

Specifications

Melting Point −10°C
Quantity 1L
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
pH 1
Color Clear
Alpha Vector Tolidine Dihydrochloride Solution
Density 1.03g/cm³
Concentration or Composition (by Analyte or Components) 0.1% w/v
Packaging Glass Bottle
MDL Number MFCD00012960
CAS 7732-18-5
ChemAlert Storage Symbol White
Sensitivity Pass Test
CAS 7647-01-0
Name Note 0.1% (w/v)
Chemical Name or Material o-Tolidine Dihydrochloride Solution
CAS 612-82-8

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

Safety and Handling