Learn More
o-Tolidine dihydrochloride, ACS, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03662622
Description
o-Tolidine is employed as a colorimetric reagent for gold and free chlorine in water. Used in the synthesis of 9,9-bis(3-methyl-4-aminophenyl)fluorine. It is utilized for the detection of reducing sugars by thin layer chromatography. It is used in hair dyes, as a pharmaceutical intermediate and laboratory reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
o-Tolidine dihydrochloride | |
>300°C | |
Odorless | |
MFCD00012960 | |
UN3077 | |
3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl | |
LUKPNZHXJRJBAN-UHFFFAOYSA-N | |
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride | |
108938 | |
ACS |
612-82-8 | |
White to Cream | |
C14H18Cl2N2 | |
100 g | |
Air and light sensitive | |
Soluble in water,alcohol,ether and dilute acids | |
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl | |
285.212 | |
285.22 | |
Solid |
Chemical Identifiers
612-82-8 | |
285.212 | |
LUKPNZHXJRJBAN-UHFFFAOYSA-N | |
108938 | |
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl |
C14H18Cl2N2 | |
MFCD00012960 | |
3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl | |
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride |
Safety and Handling
GHS H Statement
H350-H302
May cause cancer.
Harmful if swallowed.
P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H350
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 210-322-5
RTECSNumber : DD1226000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only