missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Tolidine dihydrochloride, Acros Organics™

Manufacturer:  Acros Organics AC449665000

 View more versions of this product

Catalog No. AC449665000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

o-Tolidine dihydrochloride
Authentic
500g
C14H18Cl2N2
LUKPNZHXJRJBAN-UHFFFAOYSA-N
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
108938
ACS Reagent
0.1% max.
130.0°C
612-82-8
3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl
285.212
285.2

Chemical Identifiers

612-82-8
285.212
3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
C14H18Cl2N2
LUKPNZHXJRJBAN-UHFFFAOYSA-N
108938
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl
Safety and Handling

Safety and Handling

GHS H Statement May cause cancer. Harmful if swallowed. Toxic to aquatic life with long lasting effects.

GHS P Statement Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Danger

SDS Documents