Please login to your online account to display your discounted pricing

Alfa Aesar™ O-tert-Butyl-L-tyrosine, 99%

$346.34 - $346.34

Chemical Identifiers

Chemical Name or Material O-tert-Butyl-L-tyrosine
CAS 18822-59-8
Molecular Formula C13H19NO3
Formula Weight 237.3
MDL Number MFCD00037202
Synonym H-Tyr(tBu)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2257506

 
alfa aesar™
B2257506
5g Each for $346.34
Description & Specifications

Specifications

Formula Weight 237.3
Chemical Name or Material O-tert-Butyl-L-tyrosine
Assay 99%
Molecular Formula C13H19NO3
MDL Number MFCD00037202
CAS 18822-59-8
Synonym H-Tyr(tBu)-OH
Melting Point ≈205°C (dec.)
Quantity 5g